Learn More
Medchemexpress LLC 5-Hydroxyferulic acid | 1782-55-4 | 99.4% | 210.18 | 5 MG
SDP
Catalog No. 5000364106
Encompass_Preferred
5-Hydroxyferulic acid is a hydroxycinnamic acid and a metabolite of the phenylpropanoid pathway. It serves as a precursor in the biosynthesis of sinapic acid and is also a COMT non-esterified substrate. This off-white to light yellow solid is for research use only.
  • Off-white to light yellow solid
  • Precursor in sinapic acid biosynthesis
  • COMT non-esterified substrate
  • Causes skin irritation and serious eye irritation
  • May cause respiratory irritation
  • For research use only
Catalog No. 50-003-64106 Supplier Medchemexpress LLC Supplier No. 5MGHY133068
5-Hydroxyferulic acid is a hydroxycinnamic acid and a metabolite of the phenylpropanoid pathway. It serves as a precursor in the biosynthesis of sinapic acid and is also a COMT non-esterified substrate. This off-white to light yellow solid is for research use only.
  • Off-white to light yellow solid
  • Precursor in sinapic acid biosynthesis
  • COMT non-esterified substrate
  • Causes skin irritation and serious eye irritation
  • May cause respiratory irritation
  • For research use only