Learn More
Medchemexpress LLC 5-Hydroxyflavone | 491-78-1 | 99.8% | 238.24 | 25 MG
SDP
Catalog No. 5000402292
Encompass_Preferred
5-Hydroxyflavone is a flavonoid ligand that shows no cytotoxic activity against MCF-7, FaDU, MDA-MB-435S, U87, RPE-1, and HEK293 cells.
  • Flavonoid ligand
  • No cytotoxic activity against MCF-7, FaDU, MDA-MB-435S, U87, RPE-1, and HEK293 cells
  • Solid, light yellow to yellow appearance
  • Powder storage: -20°C for 3 years, 4°C for 2 years
  • In solvent storage: -80°C for 2 years, -20°C for 1 year
Catalog No. 50-004-02292 Supplier Medchemexpress LLC Supplier No. 25MGHY22024
5-Hydroxyflavone is a flavonoid ligand that shows no cytotoxic activity against MCF-7, FaDU, MDA-MB-435S, U87, RPE-1, and HEK293 cells.
  • Flavonoid ligand
  • No cytotoxic activity against MCF-7, FaDU, MDA-MB-435S, U87, RPE-1, and HEK293 cells
  • Solid, light yellow to yellow appearance
  • Powder storage: -20°C for 3 years, 4°C for 2 years
  • In solvent storage: -80°C for 2 years, -20°C for 1 year