Learn More
Medchemexpress LLC 5-Hydroxyflavone | 491-78-1 | 99.8% | 238.24 | 50 MG
SDP
Catalog No. 5000401545
Encompass_Preferred
5-Hydroxyflavone is a flavonoid ligand that has demonstrated no cytotoxic activity against several cell lines, including MCF-7, FaDU, MDA-MB-435S, U87, RPE-1, and HEK293 cells.
  • Flavonoid ligand
  • No cytotoxic activity against various cell lines
  • Solid appearance, light yellow to yellow color
  • Soluble in DMSO
  • Long-term storage recommended at -20°C for powder and -80°C in solvent
Catalog No. 50-004-01545 Supplier Medchemexpress LLC Supplier No. HY2202450MG
5-Hydroxyflavone is a flavonoid ligand that has demonstrated no cytotoxic activity against several cell lines, including MCF-7, FaDU, MDA-MB-435S, U87, RPE-1, and HEK293 cells.
  • Flavonoid ligand
  • No cytotoxic activity against various cell lines
  • Solid appearance, light yellow to yellow color
  • Soluble in DMSO
  • Long-term storage recommended at -20°C for powder and -80°C in solvent