Learn More
Medchemexpress LLC 5-Hydroxyflavone | 491-78-1 | 99.83% | 238.24 | 1 ML
SDP
Catalog No. 5000400680
Encompass_Preferred
5-Hydroxyflavone is a flavonoid ligand that exhibits no cytotoxic activity against MCF-7, FaDU, MDA-MB-435S, U87, RPE-1, and HEK293 cells. It is intended for research use only.
  • Available in solid and solution forms.
  • Solid and solvent options available for reconstitution.
  • Powder storage: -20°C for 3 years; 4°C for 2 years.
  • In solvent storage: -80°C for 2 years; -20°C for 1 year.
  • Soluble in DMSO.
Catalog No. 50-004-00680 Supplier Medchemexpress LLC Supplier No. HY2202410MM/1ML
5-Hydroxyflavone is a flavonoid ligand that exhibits no cytotoxic activity against MCF-7, FaDU, MDA-MB-435S, U87, RPE-1, and HEK293 cells. It is intended for research use only.
  • Available in solid and solution forms.
  • Solid and solvent options available for reconstitution.
  • Powder storage: -20°C for 3 years; 4°C for 2 years.
  • In solvent storage: -80°C for 2 years; -20°C for 1 year.
  • Soluble in DMSO.