Learn More
Medchemexpress LLC HY-W008253 100mg Medchemexpress, 5-Hydroxyindole-3-acetic acid CAS:54-16-0 Purity:>98%
SDP
Catalog No. 501934509
Encompass_Preferred
Medchemexpress, HY-W008253 100mg 5-Hydroxyindole-3-acetic acid CAS:54-16-0 5-Hydroxyindole-3-acetic acid is the main metabolite of serotonin or metanephrines, which can be used as a biomarker of neuroendocrine tumors. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4509 Supplier Medchemexpress LLC Supplier No. HYW008253100MG
Medchemexpress, HY-W008253 100mg 5-Hydroxyindole-3-acetic acid CAS:54-16-0 5-Hydroxyindole-3-acetic acid is the main metabolite of serotonin or metanephrines, which can be used as a biomarker of neuroendocrine tumors. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.