Learn More
Medchemexpress LLC 5-Hydroxyindole-3-acetic acid | 54-16-0 | 191.18 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYW008253R10MG
Encompass_Preferred
5-Hydroxyindole-3-acetic acid is an analytical standard intended for research and analytical applications. It is the main metabolite of serotonin or metanephrines, which can be used as a biomarker of neuroendocrine tumors.
  • Analytical and reference standard for assay
  • Used in qualitative, quantitative, and methodological research experiments
  • Suitable for HPLC, GC, and MS applications
Catalog No. 50-004-45772