Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Hydroxyindole-3-acetic Acid, 99%, ACROS Organics™

Click to view available options
Quantity:
250mg
1g
5g
Packaging:
Glass bottle
54-16-0
C10H9NO3
191.19
MFCD00005639
DUUGKQCEGZLZNO-UHFFFAOYSA-N
5-hydroxyindole-3-acetic acid,5-hiaa,2-5-hydroxy-1h-indol-3-yl acetic acid,5-hydroxyindoleacetic acid,5-hydroxyheteroauxin,5-hydroxy-1h-indol-3-yl acetic acid,5-oxyindoleacetic acid,hydroxyindoleacetic acid,5-hydroxy-iaa,5-hydroxyindol-3-ylacetic acid
1826
CHEBI:27823
2-(5-hydroxy-1H-indol-3-yl)acetic acid
C1=CC2=C(C=C1O)C(=CN2)CC(=O)O
54-16-0
C10H9NO3
191.19
MFCD00005639
DUUGKQCEGZLZNO-UHFFFAOYSA-N
5-hydroxyindole-3-acetic acid,5-hiaa,2-5-hydroxy-1h-indol-3-yl acetic acid,5-hydroxyindoleacetic acid,5-hydroxyheteroauxin,5-hydroxy-1h-indol-3-yl acetic acid,5-oxyindoleacetic acid,hydroxyindoleacetic acid,5-hydroxy-iaa,5-hydroxyindol-3-ylacetic acid
1826
CHEBI:27823
2-(5-hydroxy-1H-indol-3-yl)acetic acid
C1=CC2=C(C=C1O)C(=CN2)CC(=O)O
Packaging Glass bottle
Water 0.5% max. (K.F.)
Quantity 250mg
Color Beige
Melting Point 162°C to 165°C
Infrared Spectrum Authentic
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Solubility Information Solubility in water: slightly soluble
Formula Weight 191.19
Physical Form Crystalline Powder
Percent Purity ≥98.5%
Chemical Name or Material 5-Hydroxyindole-3-acetic acid
Show More Show Less