Learn More
Medchemexpress LLC 5-Hydroxyindole-3-acetic acid-d6 | 197.22 | 1 MG
SDP
Catalog No. 5000445771
Encompass_Preferred
5-Hydroxyindole-3-acetic acid-d6 is the deuterium labeled form of 5-Hydroxyindole-3-acetic acid. This compound is recognized as the main metabolite of serotonin or metanephrines, serving as a biomarker for neuroendocrine tumors. Stable heavy isotopes, such as deuterium, are incorporated into drug molecules primarily as tracers for quantitation during the drug development process, and have shown potential to influence the pharmacokinetic and metabolic profiles of drugs.
  • Main metabolite of serotonin or metanephrines
  • Biomarker for neuroendocrine tumors
  • Deuterium labeled compound
  • Used as a tracer for quantitation in drug development
  • May affect pharmacokinetic and metabolic profiles
Catalog No. 50-004-45771 Supplier Medchemexpress LLC Supplier No. HYW008253S11MG
5-Hydroxyindole-3-acetic acid-d6 is the deuterium labeled form of 5-Hydroxyindole-3-acetic acid. This compound is recognized as the main metabolite of serotonin or metanephrines, serving as a biomarker for neuroendocrine tumors. Stable heavy isotopes, such as deuterium, are incorporated into drug molecules primarily as tracers for quantitation during the drug development process, and have shown potential to influence the pharmacokinetic and metabolic profiles of drugs.
  • Main metabolite of serotonin or metanephrines
  • Biomarker for neuroendocrine tumors
  • Deuterium labeled compound
  • Used as a tracer for quantitation in drug development
  • May affect pharmacokinetic and metabolic profiles