Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Hydroxyisophthalic Acid 97.0+%, TCI America™
SDP

Catalog No. H079425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H079425G 25 g
H0794500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H079425G Supplier TCI America Supplier No. H079425G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 618-83-7
Molecular Formula C8H6O5
Molecular Weight (g/mol) 182.131
MDL Number MFCD00002515
InChI Key QNVNLUSHGRBCLO-UHFFFAOYSA-N
Synonym 5-hydroxyisophthalic acid, 5-oxyisophthalic acid, isophthalic acid, 5-hydroxy, 1,3-benzenedicarboxylic acid, 5-hydroxy, 5-hydoxyisophthalic acid, 5-hydroxy-isophthalic acid, 5-hydroxy-1,3-benzenedicarboxylic acid, 5-hydroxyisosphthalic acid, isophthalic acid, 5-hydroxy-6ci,7ci,8ci, 5-hydroxyisophthalicacid
PubChem CID 69257
IUPAC Name 5-hydroxybenzene-1,3-dicarboxylic acid
SMILES C1=C(C=C(C=C1C(=O)O)O)C(=O)O

Specifications

Color White
Quantity 25 g
Formula Weight 182.13
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Hydroxyisophthalic Acid
EINECSNumber (3)-3476
RTECSNumber CZ4280000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.