Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Hydroxyisoquinoline, 90%, technical, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
2439-04-5
C9H7NO
145.16
MFCD00006906
CSNXUYRHPXGSJD-UHFFFAOYSA-N
5-hydroxyisoquinoline,5-isoquinolinol,5-hydroxy-isoquinoline,chembl57481,5-hydroxyisoquinolne,pubchem12826,acmc-2097ay,5-hydroxy-2-azanaphthalene
30386
isoquinolin-5-ol
C1=CC2=C(C=CN=C2)C(=C1)O
2439-04-5
C9H7NO
145.16
MFCD00006906
CSNXUYRHPXGSJD-UHFFFAOYSA-N
5-hydroxyisoquinoline,5-isoquinolinol,5-hydroxy-isoquinoline,chembl57481,5-hydroxyisoquinolne,pubchem12826,acmc-2097ay,5-hydroxy-2-azanaphthalene
30386
isoquinolin-5-ol
C1=CC2=C(C=CN=C2)C(=C1)O
Melting Point 22°C
Packaging Glass bottle
Color Beige
Quantity 25g
Infrared Spectrum Authentic
CAS Min % 88.0
CAS Max % 100.0
Assay Percent Range 90%
Beilstein 21,102
Solubility Information Solubility in water: soluble
Formula Weight 145.16
Physical Form Granular Powder
Percent Purity ≥88%
Grade Technical
Chemical Name or Material 5-Hydroxyisoquinoline
Show More Show Less

GHS H Statement:
Harmful in contact with skin.
Causes serious eye irritation.
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning