Learn More
TARGETMOL CHEMICALS INC 5-Hydroxymethyl-2-deoxycytidin
SDP
Catalog No. 502877191
Encompass_Preferred
Also available in 1 mL, 1 mg, 5 mg, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. 5-Hydroxymethyl-2'-deoxycytidine is an oxidized derivative of 5-methyl-2'-deoxycytidine (5-mdC) in DNA, causing DNA damage reactions, chromosomal aberrations, replication fork damage, and loss of cell viability.5-Hydroxymethyl-2'-deoxycytidine is an oxidized derivative of 5-methyl-2'-deoxycytidine in DNA. Hydroxymethyl-2'-deoxycytidine replication fork instability is associated with the presence of poly(ADP-ribose) polymerase 1 (PARP1) on chromatin. Purity 99.8%
Catalog No. 50-287-7191 Supplier TARGETMOL CHEMICALS INC Supplier No. T4083410MG
Also available in 1 mL, 1 mg, 5 mg, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. 5-Hydroxymethyl-2'-deoxycytidine is an oxidized derivative of 5-methyl-2'-deoxycytidine (5-mdC) in DNA, causing DNA damage reactions, chromosomal aberrations, replication fork damage, and loss of cell viability.5-Hydroxymethyl-2'-deoxycytidine is an oxidized derivative of 5-methyl-2'-deoxycytidine in DNA. Hydroxymethyl-2'-deoxycytidine replication fork instability is associated with the presence of poly(ADP-ribose) polymerase 1 (PARP1) on chromatin. Purity 99.8%