Learn More
Medchemexpress LLC 5-Hydroxymethyl-2'-deoxyuridine | 5116-24-5 | >99.9% | 100 MG
SDP
Catalog No. 5000338548
Encompass_Preferred
5-Hydroxymethyl-2'-deoxyuridine is a nucleoside analog that has demonstrated inhibitory effects on the replication of various human leukemia cell lines, with IC50 values ranging from 1.7-5.8 μM. It has also been shown to prolong the survival of mice afflicted with L1210 leukemia. This compound is valuable for research into cell replication and leukemia.
  • Inhibits replication of human leukemia cell lines.
  • Prolongs survival in mouse models of leukemia.
  • Useful for research on cell replication.
  • Inhibits herpes simplex virus type 1 pyrimidine 2'-deoxyribonucleoside kinase.
  • Shows dose-dependent toxicity against a human acute promyelocytic leukemia cell line.
Catalog No. 50-003-38548 Supplier Medchemexpress LLC Supplier No. HY129983100MG
5-Hydroxymethyl-2'-deoxyuridine is a nucleoside analog that has demonstrated inhibitory effects on the replication of various human leukemia cell lines, with IC50 values ranging from 1.7-5.8 μM. It has also been shown to prolong the survival of mice afflicted with L1210 leukemia. This compound is valuable for research into cell replication and leukemia.
  • Inhibits replication of human leukemia cell lines.
  • Prolongs survival in mouse models of leukemia.
  • Useful for research on cell replication.
  • Inhibits herpes simplex virus type 1 pyrimidine 2'-deoxyribonucleoside kinase.
  • Shows dose-dependent toxicity against a human acute promyelocytic leukemia cell line.