Learn More
Accela Chembio Inc 5-Hydroxymethyl-2-furaldehyde | Purity: ?98% | Mol Wt: 126.11 | 67-47-0 | MFCD00003234 | 100 g
Supplier:  Accela Chembio Inc SY010869100G
Encompass_Preferred
5-Hydroxymethyl-2-furaldehyde | Purity: ?98% | Mol Wt: 126.11 | 67-47-0 | MFCD00003234 | 100 g
Catalog No. NC1558096