Learn More
Apexbio Technology LLC 5-hydroxymethyl-2-furaldehyde 67-47-0 20mg
SDP
Catalog No. 5000087723
Encompass_Preferred
5-hydroxymethyl-2-furaldehyde (67-47-0) is a small-molecule inhibitor targeting yeast metabolic pathways It is designed to suppress yeast growth and fermentation thereby modulating microbial metabolism 5-hydroxymethyl-2-furaldehyde exerts its biological activity primarily through inhibition of yeast metabolic processes In cell-based studies 5-hydroxymethyl-2-furaldehyde demonstrates inhibitory activity with an IC50 value of approximately 2 5 5 0 mM depending on yeast strain and experimental conditions Based on these pharmacological properties 5-hydroxymethyl-2-furaldehyde holds research potential in studies of yeast biology fermentation inhibition mechanisms and microbial metabolism
Catalog No. 50-000-87723 Supplier Apexbio Technology LLC Supplier No. N133520
5-hydroxymethyl-2-furaldehyde (67-47-0) is a small-molecule inhibitor targeting yeast metabolic pathways It is designed to suppress yeast growth and fermentation thereby modulating microbial metabolism 5-hydroxymethyl-2-furaldehyde exerts its biological activity primarily through inhibition of yeast metabolic processes In cell-based studies 5-hydroxymethyl-2-furaldehyde demonstrates inhibitory activity with an IC50 value of approximately 2 5 5 0 mM depending on yeast strain and experimental conditions Based on these pharmacological properties 5-hydroxymethyl-2-furaldehyde holds research potential in studies of yeast biology fermentation inhibition mechanisms and microbial metabolism