Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 5-Hydroxymethyl-2-furancarboxylic acid | 6338-41-6 | 142.11 | 1 G
SDP

Catalog No. 5000445498
Encompass_Preferred

5-Hydroxymethyl-2-furancarboxylic acid is an endogenous metabolite of 5-hydroxymethyl-2-furfural (HMF) that is eliminated renally. It can be used as a biomarker for HMF exposure. This product is a white to yellow solid with a purity of 99.6%.

  • Main metabolite of 5-hydroxymethyl-2-furfural (HMF)
  • Valuable as a biomarker for HMF exposure
  • Target: Endogenous Metabolite
  • Pathway: Metabolic Enzyme/Protease
  • Storage as powder: -20°C for 3 years, 4°C for 2 years
  • Storage in solvent: -80°C for 6 months, -20°C for 1 month

Catalog No. 50-004-45498 Supplier Medchemexpress LLC Supplier No. HYW0052411G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

5-Hydroxymethyl-2-furancarboxylic acid is an endogenous metabolite of 5-hydroxymethyl-2-furfural (HMF) that is eliminated renally. It can be used as a biomarker for HMF exposure. This product is a white to yellow solid with a purity of 99.6%.

  • Main metabolite of 5-hydroxymethyl-2-furfural (HMF)
  • Valuable as a biomarker for HMF exposure
  • Target: Endogenous Metabolite
  • Pathway: Metabolic Enzyme/Protease
  • Storage as powder: -20°C for 3 years, 4°C for 2 years
  • Storage in solvent: -80°C for 6 months, -20°C for 1 month

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.