Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-(Hydroxymethyl)-2-iodophenol,97%, Thermo Scientific™

Catalog No. MO07809DA
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
MO07809DA 1 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MO07809DA Supplier Thermo Scientific Chemicals Supplier No. MO07809DA
Add to Cart

Chemical Identifiers

CAS 773869-57-1
Molecular Formula C7H7IO2
Molecular Weight (g/mol) 250.035
InChI Key VIWWSAGABDIIFF-UHFFFAOYSA-N
Synonym 5-hydroxymethyl-2-iodophenol, 3-hydroxy-4-iodobenzyl alcohol, 3-hydroxy-4-iodobenzenemethanol, benzenemethanol,3-hydroxy-4-iodo, zlchem 794, 5-hydroxymethyl-2-iodo-phenol, 3-hydroxy-4-iodophenyl methanol, 5-hydroxymethyl-2-iodanyl-phenol
PubChem CID 18525941
IUPAC Name 5-(hydroxymethyl)-2-iodophenol
SMILES C1=CC(=C(C=C1CO)O)I

Specifications

Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 250.03
Percent Purity 97%
Chemical Name or Material 5-(hydroxymethyl)-2-iodophenol
Hazard Category GHS Signal Word: Warning
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.