Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Hydroxymethyl-5-methyl-1,3-dioxane 97%, Thermo Scientific™

Catalog No. AC136021000
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC136021000 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC136021000 Supplier Thermo Scientific Chemicals Supplier No. 136021000

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1121-97-7
Molecular Formula C6H12O3
Molecular Weight (g/mol) 132.16
MDL Number MFCD00023825
InChI Key XZDDZTGNCUMJHI-UHFFFAOYSA-N
Synonym 5-hydroxymethyl-5-methyl-1,3-dioxane, 5-methyl-1,3-dioxan-5-yl methanol, 5-methyl-1,3-dioxane-5-methanol, m-dioxane-5-methanol, 5-methyl, 1,3-dioxane-5-methanol, 5-methyl, 1,3-dioxane-5-methanol,5-methyl, 5-methyl-1,3-dioxan-5-yl methan-1-ol, 5-19-02-00358 beilstein handbook reference, 5-methyl-m-dioxane-5-methanol
PubChem CID 14278
IUPAC Name (5-methyl-1,3-dioxan-5-yl)methanol
SMILES CC1(CO)COCOC1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Colorless
Density 1.12g/mL
Infrared Spectrum Authentic
Assay Percent Range 97%
Refractive Index 1.4565 to 1.4585 (20°C, 589nm)
Quantity 100 g
Formula Weight 132.16
Percent Purity ≥96.0%
Physical Form Viscous Liquid
Chemical Name or Material 5-Hydroxymethyl-5-methyl-1, 3-dioxane, 97%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.