Learn More
Medchemexpress LLC 5-hydroxymethyl uridine | 30414-00-7 | MFCD00057401 | ≥98.0% | 274.23 g/mol | C10H14N2O7 | 1 ML
SDP
Catalog No. 5000281489
Encompass_Preferred
5-Hydroxymethyl uridine is a thymidine analogue nucleoside used to label newly synthesized DNA and monitor cell proliferation. It is supplied as a high-purity reagent in solid form or as ready-to-use solutions (for example, 10 mM in DMSO), and is suitable for biochemical and cell-based assays when stored and handled under recommended conditions.
  • Thymidine analogue for incorporation into newly synthesized DNA.
  • High purity (≥98.0%) suitable for analytical and biological applications.
  • Available as solid or as a 10 mM solution in DMSO for flexible workflows.
  • Soluble in common laboratory solvents for convenient preparation of working solutions.
  • Stable under recommended storage conditions for reproducible results.
  • Suitable for cell labeling and tracking DNA synthesis in proliferation assays.
Catalog No. 50-002-81489 Supplier Medchemexpress LLC Supplier No. HYW39410610MM/1ML
5-Hydroxymethyl uridine is a thymidine analogue nucleoside used to label newly synthesized DNA and monitor cell proliferation. It is supplied as a high-purity reagent in solid form or as ready-to-use solutions (for example, 10 mM in DMSO), and is suitable for biochemical and cell-based assays when stored and handled under recommended conditions.
  • Thymidine analogue for incorporation into newly synthesized DNA.
  • High purity (≥98.0%) suitable for analytical and biological applications.
  • Available as solid or as a 10 mM solution in DMSO for flexible workflows.
  • Soluble in common laboratory solvents for convenient preparation of working solutions.
  • Stable under recommended storage conditions for reproducible results.
  • Suitable for cell labeling and tracking DNA synthesis in proliferation assays.