Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 5-hydroxymethyl uridine | 30414-00-7 | MFCD00057401 | 98.0% | 274.23 g/mol | C10H14N2O7 | 100 MG
SDP

Catalog No. 5000281488
Encompass_Preferred

5-Hydroxymethyl uridine is a thymidine analogue used to label and track DNA synthesis in replicating cells. It incorporates into DNA during replication and is suitable for biochemical assays and cell-labeling applications that require nucleoside analogs.

  • Incorporates into replicating DNA for labeling and detection.
  • High purity (98.0%) suitable for biochemical assays.
  • Solid, white to light yellow appearance for easy handling.
  • Available in 100 MG powder and other pack sizes.
  • Molecular weight 274.23 g/mol; formula C10H14N2O7.
  • Recommended storage: powder -20°C (up to 3 years) or 4°C (up to 2 years).

Catalog No. 50-002-81488 Supplier Medchemexpress LLC Supplier No. HYW394106100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

5-Hydroxymethyl uridine is a thymidine analogue used to label and track DNA synthesis in replicating cells. It incorporates into DNA during replication and is suitable for biochemical assays and cell-labeling applications that require nucleoside analogs.

  • Incorporates into replicating DNA for labeling and detection.
  • High purity (98.0%) suitable for biochemical assays.
  • Solid, white to light yellow appearance for easy handling.
  • Available in 100 MG powder and other pack sizes.
  • Molecular weight 274.23 g/mol; formula C10H14N2O7.
  • Recommended storage: powder -20°C (up to 3 years) or 4°C (up to 2 years).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.