Learn More
Cayman Chemical 5-(Hydroxymethyl)-2’-deoxycytidine, Purity: ≥95%, CAS: 7226-77-9, 10 mg
SDP
Catalog No. 502966795
Encompass_Preferred
A modified pyrimidine that is capable of producing interstrand cross-links in double-stranded DNA; used to quantify DNA hydroxymethylation levels in biological samples
Catalog No. 50-296-6795 Supplier Cayman Chemical Supplier No. 1816210 MG
A modified pyrimidine that is capable of producing interstrand cross-links in double-stranded DNA; used to quantify DNA hydroxymethylation levels in biological samples