Learn More
Cayman Chemical 5-(Hydroxymethyl)-2’-deoxyuridine, Purity: ≥98%, CAS: 5116-24-5, 100 mg
SDP
Catalog No. 502993747
Encompass_Preferred
A nucleoside analog; inhibits the replication of murine S180 lung carcinoma cells and Ehrlich ascites mammary carcinoma cells (ED50s = 8.5 and 4 μM, respectively) and multiple human leukemia cell lines (IC50s = 1.7-5.8 μM); acts synergistically with 5-FU against HT-29, HCT116, PANC-1, and EKVX cancer cells; inhibits HSV-1 pyrimidine 2’-deoxyribonucleoside kinase (Ki = 3.5 μM) and reduces HSV-1 viral titer to 0.05% of the control at a concentration of 200 μM; a DNA adduct formed in response to oxidative stress
Catalog No. 50-299-3747 Supplier Cayman Chemical Supplier No. 23381100 MG
A nucleoside analog; inhibits the replication of murine S180 lung carcinoma cells and Ehrlich ascites mammary carcinoma cells (ED50s = 8.5 and 4 μM, respectively) and multiple human leukemia cell lines (IC50s = 1.7-5.8 μM); acts synergistically with 5-FU against HT-29, HCT116, PANC-1, and EKVX cancer cells; inhibits HSV-1 pyrimidine 2’-deoxyribonucleoside kinase (Ki = 3.5 μM) and reduces HSV-1 viral titer to 0.05% of the control at a concentration of 200 μM; a DNA adduct formed in response to oxidative stress