Learn More
Cayman Chemical 5-HydroxymethylcytIdIn 10mg
SDP
Catalog No. 502945411
Encompass_Preferred
A derivative of cytidine; has been found as an RNA modification in Archaea, bacteria, and eukaryotes
Catalog No. 50-294-5411 Supplier Cayman Chemical Supplier No. 4074510 MG
A derivative of cytidine; has been found as an RNA modification in Archaea, bacteria, and eukaryotes