Learn More
Epigentek Inc 5-Hydroxymethylcytosine (5-hmC) Monoclonal Antibody [HMC/4D9] (50 ug)
SDP
Supplier:  Epigentek Inc A1018050
Encompass_Preferred
Unconjugated Monoclonal Antibody Raised in Mouse. Immunogen: . Tested Applications: IF, IHC, ELISA, DB, MeDIP. Reactivity: Broad Range. Alternative Names: 5-hydroxymethylcytidine, anti-5-hydroxymethylcytidine, anti-5-hydroxymethylcytosine, anti-5hmC, anti-5-hmC, anti-5-hmeC, anti-5hmeC, 5hmC, 5hmeC, 5-hmeC.
Catalog No. 50-109-4958