Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Matrix Scientific 5-Hydroxymethylfurfural, 67-47-0, MFCD00003234, 5g

Catalog No. 50418686
Encompass_Preferred

Molecular Formula C6H6O3, Purity 98+%, Molecular Weight 126.11, Melting Point 32-35°, Boiling Point 114-116/1mm

Catalog No. 50-418-686 Supplier Matrix Scientific Supplier No. 0729255G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Molecular Formula C6H6O3, Purity 98+%, Molecular Weight 126.11, Melting Point 32-35°, Boiling Point 114-116/1mm

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.