Learn More
Medchemexpress LLC HY-W004924 1g Medchemexpress, 5-Hydroxymethyluracil CAS:4433-40-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYW0049241G
Encompass_Preferred
Medchemexpress, HY-W004924 1g 5-Hydroxymethyluracil CAS:4433-40-3 5-Hydroxymethyluracil is a product of oxidative DNA damage. 5-Hydroxymethyluracil can be used as a potential epigenetic mark enhancing or inhibiting transcription with bacterial RNA polymerase. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4489