Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Hydroxyoxindole, 96%, Thermo Scientific Chemicals

Catalog No. AC153550010
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC153550010 1 g
AC153550050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC153550010 Supplier Thermo Scientific Chemicals Supplier No. 153550010
Add to Cart

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3416-18-0
Molecular Formula C8H7NO2
Molecular Weight (g/mol) 149.15
InChI Key ZGTUSQAQXWSMDW-UHFFFAOYSA-N
Synonym 5-hydroxyoxindole, 5-hydroxyindolin-2-one, 5-hydroxy-1,3-dihydro-indol-2-one, 2,3-dihydro-5-hydroxyindol-2-one, 5-hydroxy-2-oxyindole, 5-hydroxy-2-indolinone, 5-hydroxy-1,3-dihydro-2h-indol-2-one, 5-hydroxy-2,3-dihydro-1h-indol-2-one, 2-indolinone, 5-hydroxy, 5-hydroxy-oxindole
PubChem CID 76955
IUPAC Name 5-hydroxy-1,3-dihydroindol-2-one
SMILES C1C2=C(C=CC(=C2)O)NC1=O

Specifications

Melting Point 266.0°C to 272.0°C
Assay Percent Range 95% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 149.15
Percent Purity 96%
Chemical Name or Material 5-Hydroxyoxindole
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
May cause an allergic skin reaction.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritatio
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and kee
EINECSNumber 222-309-1

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.