Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Hydroxypentanal (Tech.), Thermo Scientific™

Catalog No. AC156410250
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC156410250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC156410250 Supplier Thermo Scientific Chemicals Supplier No. 156410250

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4228-10-8
Molecular Formula C11H12O
Molecular Weight (g/mol) 160.22
InChI Key HCMWPTFPUCPKQM-UHFFFAOYSA-N
Synonym 5-acetylindane, 5-acetylindan, 1-2,3-dihydro-1h-inden-5-yl ethanone, 1-2,3-dihydro-1h-inden-5-yl ethan-1-one, ethanone,3-dihydro-1h-inden-5-yl, indan-5-ylethan-1-one, 1-indane-5-yl-ethanone, acmc-1arc8, 1-2,3-dihydro-1h-inden-6-yl ethanone, 1-2,3-dihydro-1h-inden-5-yl-ethanone
PubChem CID 233976
IUPAC Name 1-(2,3-dihydro-1H-inden-5-yl)ethanone
SMILES CC(=O)C1=CC2=C(CCC2)C=C1

Specifications

CAS Min % 88.0
CAS Max % 100.0
Color Colorless to Yellow
Density 1.05
Infrared Spectrum Authentic
Linear Formula HO(CH2)4CHO
Quantity 25 g
Formula Weight 102.13
Percent Purity ≥88%
Grade Technical
Physical Form Liquid
Chemical Name or Material 5-Hydroxypentanal, tech.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.