Learn More
Medchemexpress LLC 5-hydroxypyrazine-2-carboxylic acid | 34604-60-9 | 99.99% | C5H4N2O3 | 50 G
SDP
Catalog No. 5000434676
Encompass_Preferred
5-Hydroxypyrazine-2-Carboxylic Acid is a metabolite of the anti-tuberculosis drug pyrazinamide (PZA). Supplied as a solid, this product is intended for research use only.
  • Metabolite of the anti-tuberculosis drug pyrazinamide (PZA)
  • For research use only
  • Supplied as a solid
  • Purity of 99.99%
  • Stable under recommended storage conditions
Catalog No. 50-004-34676 Supplier Medchemexpress LLC Supplier No. HY7621050G
5-Hydroxypyrazine-2-Carboxylic Acid is a metabolite of the anti-tuberculosis drug pyrazinamide (PZA). Supplied as a solid, this product is intended for research use only.
  • Metabolite of the anti-tuberculosis drug pyrazinamide (PZA)
  • For research use only
  • Supplied as a solid
  • Purity of 99.99%
  • Stable under recommended storage conditions