Learn More
Medchemexpress LLC 5-Hydroxyquinoline | 578-67-6 | 98.4% | 145.16 | 10 G
SDP
Catalog No. 5000401987
Encompass_Preferred
5-Hydroxyquinoline is an organic compound commonly used in certain chemical reactions and biological research. It can be used in the manufacture of dyes, cellulase, oxidants and passivators, etc., and is widely used in the pigment, paint and rubber industries. In addition, the compound is also used as the precursor, intermediate and metal ion complexing agent of certain drugs.
  • Is a biochemical reagent that can be used as a biological material or organic compound for life science related research.
Catalog No. 50-004-01987 Supplier Medchemexpress LLC Supplier No. HY3210010G
5-Hydroxyquinoline is an organic compound commonly used in certain chemical reactions and biological research. It can be used in the manufacture of dyes, cellulase, oxidants and passivators, etc., and is widely used in the pigment, paint and rubber industries. In addition, the compound is also used as the precursor, intermediate and metal ion complexing agent of certain drugs.
  • Is a biochemical reagent that can be used as a biological material or organic compound for life science related research.