Learn More
Medchemexpress LLC 5-Hydroxyquinoline | 578-67-6 | 99.5% | 145.16 | 1 G
SDP
Catalog No. 5000401230
Encompass_Preferred
5-Hydroxyquinoline is an organic compound commonly used in chemical reactions and biological research. It is utilized in the manufacture of dyes, cellulase, oxidants, and passivators, and finds wide application in the pigment, paint, and rubber industries. This compound also serves as a precursor, intermediate, and metal ion complexing agent for various drugs, and is a biochemical reagent for life science-related research.
  • Used in chemical reactions and biological research.
  • Manufacture of dyes, cellulase, oxidants, and passivators.
  • Widely used in pigment, paint, and rubber industries.
  • Precursor, intermediate, and metal ion complexing agent for certain drugs.
  • Biochemical reagent for life science related research.
  • Laboratory chemical.
  • For research use only.
Catalog No. 50-004-01230 Supplier Medchemexpress LLC Supplier No. HY321001G
5-Hydroxyquinoline is an organic compound commonly used in chemical reactions and biological research. It is utilized in the manufacture of dyes, cellulase, oxidants, and passivators, and finds wide application in the pigment, paint, and rubber industries. This compound also serves as a precursor, intermediate, and metal ion complexing agent for various drugs, and is a biochemical reagent for life science-related research.
  • Used in chemical reactions and biological research.
  • Manufacture of dyes, cellulase, oxidants, and passivators.
  • Widely used in pigment, paint, and rubber industries.
  • Precursor, intermediate, and metal ion complexing agent for certain drugs.
  • Biochemical reagent for life science related research.
  • Laboratory chemical.
  • For research use only.