Learn More
Medchemexpress LLC 5-Hydroxyquinoline | 578-67-6 | 99.51% | 145.16 | 50 G
SDP
Catalog No. 5000400653
Encompass_Preferred
5-Hydroxyquinoline is an organic compound commonly used in certain chemical reactions and biological research. It can be used in the manufacture of dyes, cellulase, oxidants and passivators, etc., and is widely used in the pigment, paint and rubber industries. Additionally, the compound is utilized as a precursor, intermediate, and metal ion complexing agent for specific drugs. It is a biochemical reagent that can be used as a biological material or organic compound for life science related research.
  • Used in chemical reactions and biological research.
  • Utilized in the manufacture of dyes, cellulase, oxidants and passivators.
  • Widely applied in pigment, paint, and rubber industries.
  • Functions as a precursor, intermediate, and metal ion complexing agent for specific drugs.
  • Biochemical reagent for life science-related research.
Catalog No. 50-004-00653 Supplier Medchemexpress LLC Supplier No. HY3210050G
5-Hydroxyquinoline is an organic compound commonly used in certain chemical reactions and biological research. It can be used in the manufacture of dyes, cellulase, oxidants and passivators, etc., and is widely used in the pigment, paint and rubber industries. Additionally, the compound is utilized as a precursor, intermediate, and metal ion complexing agent for specific drugs. It is a biochemical reagent that can be used as a biological material or organic compound for life science related research.
  • Used in chemical reactions and biological research.
  • Utilized in the manufacture of dyes, cellulase, oxidants and passivators.
  • Widely applied in pigment, paint, and rubber industries.
  • Functions as a precursor, intermediate, and metal ion complexing agent for specific drugs.
  • Biochemical reagent for life science-related research.