Learn More
New England Biolabs, Inc. 5-hydroxythymidine DNA Kinase – 1000 units
SDP
Supplier:  New England Biolabs, Inc. M0659S
Encompass
5-hydroxymethyluridine DNA Kinase (5-HMUDK) transfers the gamma phosphate from ATP to the hydroxymethyl moiety of 5-hydroxymethyluridine in polymeric DNA.
Catalog No. 50-202-4042