Learn More
Sigma Aldrich Fine Chemicals Biosciences Serotonin hydrochloride, 153-98-0, MFCD00012686, 25 mg
Supplier:  Sigma Aldrich Fine Chemicals Biosciences H952325MG
Encompass
Empirical Formula (Hill Notation): C10H12N2O · HCl, Molecular Weight: 212.68, powder, ≥98%, mp: 149-154 °C, Synonym: 3-(2-Aminoethyl)-5-hydroxyindole hydrochloride, 5-HT, 5-Hydroxytryptamine hydrochloride.
Catalog No. NC1298582