Learn More
Medchemexpress LLC 5-Hydroxytryptamine creatinine sulfate monohydrate | 61-47-2 | 99.9% | 405.43 | 1 ML
SDP
Catalog No. 5000366284
Encompass_Preferred
5-Hydroxytryptamine creatinine sulfate monohydrate is an endogenous metabolite. This product is a 10 mM solution in 1 mL of DMSO, suitable for various research applications.
  • Purity of 99.87%
  • Endogenous metabolite
  • Solution in DMSO
  • For research use only
Catalog No. 50-003-66284 Supplier Medchemexpress LLC Supplier No. HYW01097310MM/1ML
5-Hydroxytryptamine creatinine sulfate monohydrate is an endogenous metabolite. This product is a 10 mM solution in 1 mL of DMSO, suitable for various research applications.
  • Purity of 99.87%
  • Endogenous metabolite
  • Solution in DMSO
  • For research use only