Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 5-hydroxytryptamine-d4 | 58264-95-2 | 99.6% | 180.24 | C10H8D4N2O | 10 MG
SDP

Catalog No. 5000298953
Encompass_Preferred

Serotonin-d4 is a deuterated analogue of serotonin used as a stable isotope internal standard for quantitative mass spectrometry and tracer studies. It provides a predictable mass shift for accurate quantitation and method validation in biological and biochemical analyses.

  • Purity: 99.6%.
  • Molecular weight: 180.24 g/mol.
  • Chemical formula: C10H8D4N2O.
  • Physical form: solid, light brown to brown.
  • Solubility: DMSO 100 mg/mL; ultrasonic assistance recommended.
  • Storage: 4°C under nitrogen; in solvent -80°C up to 6 months, -20°C up to 1 month.

Catalog No. 50-002-98953 Supplier Medchemexpress LLC Supplier No. HYB1473AS10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Serotonin-d4 is a deuterated analogue of serotonin used as a stable isotope internal standard for quantitative mass spectrometry and tracer studies. It provides a predictable mass shift for accurate quantitation and method validation in biological and biochemical analyses.

  • Purity: 99.6%.
  • Molecular weight: 180.24 g/mol.
  • Chemical formula: C10H8D4N2O.
  • Physical form: solid, light brown to brown.
  • Solubility: DMSO 100 mg/mL; ultrasonic assistance recommended.
  • Storage: 4°C under nitrogen; in solvent -80°C up to 6 months, -20°C up to 1 month.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.