Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Hydroxytryptamine, MP Biomedicals™

Catalog No. ICN15376591 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
25 mg
50 mg
100 mg
250 mg
1 g
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15376591 1 g
ICN15376550 50 mg
ICN15376580 100 mg
ICN15376583 250 mg
ICN15376525 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. ICN15376591 Supplier MP Biomedicals, Inc Supplier No. 0215376591
Add to Cart
Add to Cart

Chemical Identifiers

CAS 153-98-0
Molecular Formula C10H13ClN2O
Molecular Weight (g/mol) 212.677
InChI Key MDIGAZPGKJFIAH-UHFFFAOYSA-N
Synonym serotonin hydrochloride, 5-hydroxytryptamine hydrochloride, 3-2-aminoethyl-1h-indol-5-ol hydrochloride, serotonin hcl, 5-hydroxytryptamine hcl, unii-gkn429m9vs, ccris 4420, 1h-indol-5-ol, 3-2-aminoethyl-, monohydrochloride, gkn429m9vs, 3-2-aminoethyl indol-5-ol, chloride
PubChem CID 160436
IUPAC Name 3-(2-aminoethyl)-1H-indol-5-ol;hydrochloride
SMILES C1=CC2=C(C=C1O)C(=CN2)CCN.Cl

Specifications

Color Gray
Assay Percent Range 0.993
Quantity 1 g
Formula Weight 212.7
Physical Form Powder
Chemical Name or Material 5-Hydroxytryptamine
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.