Learn More
Selleck Chemical LLC 5-hydroxytryptophan (5-HTP) S2374-50mg
SDP
Catalog No. 503112798
Encompass_Preferred
5-Hydroxytryptophan (5-HTP NSC-92523) also known as oxitriptan (INN) is a naturally occurring amino acid and chemical precursor for the treatment of depression
Catalog No. 50-311-2798 Supplier Selleck Chemical LLC Supplier No. S237450MG
5-Hydroxytryptophan (5-HTP NSC-92523) also known as oxitriptan (INN) is a naturally occurring amino acid and chemical precursor for the treatment of depression