Learn More
Cayman Chemical 5-IAIhydrochlorIde 10mg
SDP
Catalog No. 502951282
Encompass_Preferred
A psychoactive analog of p-iodoamphetamine that is indistinguishable, in its physiological effects, from MDMA in rats; significantly reduces both serotonin uptake sites and hippocampal serotonin levels in rats; intended for forensic applications
Catalog No. 50-295-1282 Supplier Cayman Chemical Supplier No. 1103510 MG
A psychoactive analog of p-iodoamphetamine that is indistinguishable, in its physiological effects, from MDMA in rats; significantly reduces both serotonin uptake sites and hippocampal serotonin levels in rats; intended for forensic applications