Learn More
Lovibond™ 5-in-1 Multi Pool Test Kit
Catalog No. 09051507 Shop All Tintometer Group Products
Change view
Click to view available options
Volume (Metric):
30 mL
Catalog No. Volume (Metric)
09-051-507 30 mL
1 options
Catalog No. 09-051-507 Supplier Lovibond™ Supplier No. 151900
Add to Cart
Add to Cart
This kit comes with everything needed to measure Alkalinity-M, Calcium Hardness, Chlorine, Cyanuric acid, and pH in pool and spa water.
  • Submerse the pool tester in water, add a rapidly-dissolving tablet, and compare the color change to the chart printed on the pool tester
  • Alkalinity: up to 20 mg/L
  • Calcium Hardness: 20-800 mg/L
  • Chlorine: 0.1 - 3.0 mg/L
  • Cyanuric acid: 20 - 200 mg/L
  • pH: 6.8 - 8.2
  • Includes a Pool tester block, measuring cylinder, two sample tubes, brush, stirring rod, instruction manual, 20x DPD No.1 Free Chlorine Rapid Tablets, 20x DPD No.3 Total Chlorine Rapid Tablets, 20x Phenol Red Rapid Tablets, 20x CyA-Test Tablets, 10x Alk LR Tablets, 10x CAL-test Tablets
Specifications
pH Range 6.8 to 8.2
Volume (Metric) 30 mL
Type Pool Tester