Learn More
Apexbio Technology LLC 5-Indolyl-AA-dUTP(Synonyms: 5-Indolyl-AA-dUTP, 5-Indolyl aminoallyl-deoxyuridine triphosphate, Aminoallyl-dUTP-5-indolyl), 100ul (10 mM).
SDP
Catalog No. 5000060747
Encompass_Preferred
5-Indolyl-AA-dUTP is a synthetic nucleotide analog designed to competitively interact with specific enzyme targets modulating DNA replication processes Through selective interaction this compound influences distinct signaling pathways implicated in cellular proliferation and differentiation Due to its ability to regulate gene expression pathways 5-Indolyl-AA-dUTP holds relevance for exploring genetic disease mechanisms and investigating potential anticancer therapeutic strategies
Catalog No. 50-000-60747 Supplier Apexbio Technology LLC Supplier No. B81180.1
5-Indolyl-AA-dUTP is a synthetic nucleotide analog designed to competitively interact with specific enzyme targets modulating DNA replication processes Through selective interaction this compound influences distinct signaling pathways implicated in cellular proliferation and differentiation Due to its ability to regulate gene expression pathways 5-Indolyl-AA-dUTP holds relevance for exploring genetic disease mechanisms and investigating potential anticancer therapeutic strategies