Learn More
Cayman Chemical 5-Iodo-2'-deoxycytidine, Purity: ≥98%, CAS: 611-53-0, 5 g
SDP
Catalog No. 502911138
Encompass_Preferred
A nucleoside analog; inhibits the growth of P815 cells (IC50 = 14 µM), as well as sensitizes P815 cells to irradiation when used at a concentration of 1 µM
Catalog No. 50-291-1138 Supplier Cayman Chemical Supplier No. 356965 G
A nucleoside analog; inhibits the growth of P815 cells (IC50 = 14 µM), as well as sensitizes P815 cells to irradiation when used at a concentration of 1 µM