Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Iodo-2'-Deoxycytidine, MP Biomedicals™

Catalog No. ICN10206201 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10206201 1 g
ICN10206205 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN10206201 Supplier MP Biomedicals, Inc Supplier No. 0210206201
Add to Cart
Add to Cart

Chemical Identifiers

CAS 611-53-0
Molecular Formula C9H12IN3O4
Molecular Weight (g/mol) 353.12
MDL Number MFCD00038063
InChI Key WEVJJMPVVFNAHZ-RRKCRQDMSA-N
Synonym 5-iodo-2'-deoxycytidine, ibacitabine, 2'-deoxy-5-iodocytidine, 4-amino-1-2r,4s,5r-4-hydroxy-5-hydroxymethyl tetrahydrofuran-2-yl-5-iodopyrimidin-2 1h-one, ibacitabine inn, ibacitabinum latin, ibacitabina spanish, cytidine, 2'-deoxy-5-iodo, iododeoxycytidine, unii-3ek8532dzv
PubChem CID 65050
IUPAC Name 4-amino-1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-5-iodo-1,2-dihydropyrimidin-2-one
SMILES NC1=NC(=O)N(C=C1I)[C@H]1C[C@H](O)[C@@H](CO)O1

Specifications

Quantity 1 g
Formula Weight 353.1
Physical Form Solid
Chemical Name or Material 5-Iodo-2'-Deoxycytidine
EINECSNumber 210-269-8
Recommended Storage Store at 0°C.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.