Learn More
Medchemexpress LLC 5-Iodo-A-85380 dihyd 5mg | 1217837-17-6 | 5MG
SDP
Catalog No. 5000242095
Encompass_Preferred
5-Iodo-A-85380 dihydrochloride is a selective ligand of nAChR 5-Iodo-A-85380 dihydrochloride binds to 4 2 nAChRs in rat and human brain with Kds of 12 and 14 pM respectively[1
Catalog No. 50-002-42095 Supplier Medchemexpress LLC Supplier No. HY1076865MG
5-Iodo-A-85380 dihydrochloride is a selective ligand of nAChR 5-Iodo-A-85380 dihydrochloride binds to 4 2 nAChRs in rat and human brain with Kds of 12 and 14 pM respectively[1