Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 5-iodo-A-85380 dihydrochloride | 1217837-17-6 | 99.0% | 363.02 | C9H13Cl2IN2O | 100 MG
SDP

Catalog No. 5000242096
Encompass_Preferred

5-Iodo-A-85380 dihydrochloride is a high-affinity, selective ligand for neuronal nicotinic acetylcholine receptors, with pronounced activity at the α4β2 subtype. Supplied as the dihydrochloride salt, it is intended for receptor binding, pharmacological profiling, and preclinical research applications. Reported binding affinities are 12 pM (rat) and 14 pM (human).

  • High affinity for α4β2 nAChR (Kd 12 pM rat, 14 pM human).
  • Provided as a dihydrochloride salt for stability in assays.
  • Purity approximately 99.0% (HPLC).
  • Molecular weight 363.02 and formula C9H13Cl2IN2O.
  • Intended for receptor binding and pharmacological characterization in preclinical studies.

Catalog No. 50-002-42096 Supplier Medchemexpress LLC Supplier No. HY107686100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

5-Iodo-A-85380 dihydrochloride is a high-affinity, selective ligand for neuronal nicotinic acetylcholine receptors, with pronounced activity at the α4β2 subtype. Supplied as the dihydrochloride salt, it is intended for receptor binding, pharmacological profiling, and preclinical research applications. Reported binding affinities are 12 pM (rat) and 14 pM (human).

  • High affinity for α4β2 nAChR (Kd 12 pM rat, 14 pM human).
  • Provided as a dihydrochloride salt for stability in assays.
  • Purity approximately 99.0% (HPLC).
  • Molecular weight 363.02 and formula C9H13Cl2IN2O.
  • Intended for receptor binding and pharmacological characterization in preclinical studies.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.