Learn More
Medchemexpress LLC 5-iodo-A-85380 dihydrochloride hydrate | 1217837-17-6 | MFCD20488048 | 99.0% | 363.02 g/mol | C9H13Cl2IN2O | 50 MG
SDP
Catalog No. 5000241319
Encompass_Preferred
5-Iodo-A-85380 dihydrochloride is a research-grade, high-affinity ligand selective for α4β2 nicotinic acetylcholine receptors. Supplied as the dihydrochloride hydrate salt, it is intended for in vitro receptor binding and pharmacology studies where subtype-selective binding is required.
  • Selective ligand for α4β2 and α6β2 nicotinic acetylcholine receptors.
  • Very high binding affinity with reported Kd values of 12 pM (rat) and 14 pM (human).
  • Supplied as the dihydrochloride hydrate salt for improved handling and stability.
  • High purity (98.97%) suitable for biochemical assays.
  • Molecular weight 363.02 g/mol; appropriate for molarity calculations in assays.
Catalog No. 50-002-41319 Supplier Medchemexpress LLC Supplier No. HY10768650MG
5-Iodo-A-85380 dihydrochloride is a research-grade, high-affinity ligand selective for α4β2 nicotinic acetylcholine receptors. Supplied as the dihydrochloride hydrate salt, it is intended for in vitro receptor binding and pharmacology studies where subtype-selective binding is required.
  • Selective ligand for α4β2 and α6β2 nicotinic acetylcholine receptors.
  • Very high binding affinity with reported Kd values of 12 pM (rat) and 14 pM (human).
  • Supplied as the dihydrochloride hydrate salt for improved handling and stability.
  • High purity (98.97%) suitable for biochemical assays.
  • Molecular weight 363.02 g/mol; appropriate for molarity calculations in assays.