Learn More
Apexbio Technology LLC 5-Iodo-CTP(Synonyms: 5-Iodocytidine-5'-triphosphate, 5-Iodo-CTP sodium salt, 5-Iodo-cytidine triphosphate), 50ul (100 mM).
SDP
Catalog No. 5000061236
Encompass_Preferred
5-Iodo-CTP is a nucleotide analog derived from cytidine triphosphate (CTP) Due to its triphosphate structure it serves as a substrate for RNA polymerases and becomes incorporated into nascent RNA strands during transcription Incorporation of the iodine atom into RNA molecules facilitates their subsequent detection and analysis Thus 5-Iodo-CTP finds utility in studying RNA structure-function relationships analyzing RNA synthesis and stability and characterizing RNA-protein interactions Additionally the iodine substituent integrated within RNA offers a convenient handle for cross-linking or probing RNA-associated molecular events
Catalog No. 50-000-61236 Supplier Apexbio Technology LLC Supplier No. B79640.05
5-Iodo-CTP is a nucleotide analog derived from cytidine triphosphate (CTP) Due to its triphosphate structure it serves as a substrate for RNA polymerases and becomes incorporated into nascent RNA strands during transcription Incorporation of the iodine atom into RNA molecules facilitates their subsequent detection and analysis Thus 5-Iodo-CTP finds utility in studying RNA structure-function relationships analyzing RNA synthesis and stability and characterizing RNA-protein interactions Additionally the iodine substituent integrated within RNA offers a convenient handle for cross-linking or probing RNA-associated molecular events