Learn More
Apexbio Technology LLC 5-Iodo-dCTP(Synonyms: 5-Iodo-2'-deoxycytidine 5'-triphosphate, 5-Iodo-dCTP, 5-Iodo-deoxycytidine triphosphate), 50ul (100 mM).
SDP
Catalog No. 5000060705
Encompass_Preferred
5-Iodo-dCTP is a bioactive deoxynucleotide analog known to modulate nucleotide metabolism by interfering with specific aspects of DNA synthesis Through incorporation into nucleic acids it disrupts normal DNA replication processes allowing detailed exploration of nucleotide biosynthesis pathways Experimentally 5-Iodo-dCTP serves as a molecular tool to probe polymerase enzyme function and analyze mechanisms underlying DNA synthesis fidelity It is commonly employed in molecular biology investigations facilitating studies on disease-associated disruptions in nucleic acid metabolism
Catalog No. 50-000-60705 Supplier Apexbio Technology LLC Supplier No. B80910.05
5-Iodo-dCTP is a bioactive deoxynucleotide analog known to modulate nucleotide metabolism by interfering with specific aspects of DNA synthesis Through incorporation into nucleic acids it disrupts normal DNA replication processes allowing detailed exploration of nucleotide biosynthesis pathways Experimentally 5-Iodo-dCTP serves as a molecular tool to probe polymerase enzyme function and analyze mechanisms underlying DNA synthesis fidelity It is commonly employed in molecular biology investigations facilitating studies on disease-associated disruptions in nucleic acid metabolism