Learn More
Medchemexpress LLC HY-111641 10mg Medchemexpress, 3'-Azido-3'-deoxy-5-fluorocytidine CAS:2095417-18-6 Purity:>98%
SDP
Catalog No. 501968932
Encompass_Preferred
Medchemexpress, HY-111930 10mg 5-Iodo-indirubin-3'-monoxime CAS:331467-03-9 5-Iodo-indirubin-3'-monoxime is a potent GSK-3β, CDK5/P25 and CDK1/cyclin B inhibitor, competing with ATP for binding to the catalytic site of the kinase, with IC50s of 9, 20 and 25 nM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8932 Supplier Medchemexpress LLC Supplier No. HY11164110MG
Medchemexpress, HY-111930 10mg 5-Iodo-indirubin-3'-monoxime CAS:331467-03-9 5-Iodo-indirubin-3'-monoxime is a potent GSK-3β, CDK5/P25 and CDK1/cyclin B inhibitor, competing with ATP for binding to the catalytic site of the kinase, with IC50s of 9, 20 and 25 nM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.