Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Iodo-m-xylene 98.0+%, TCI America™
SDP

Catalog No. I054825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I054825G 25 g
I05485G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I054825G Supplier TCI America Supplier No. I054825G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 22445-41-6 | C8H9I | 232.06 g/mol

Chemical Identifiers

CAS 22445-41-6
Molecular Formula C8H9I
Molecular Weight (g/mol) 232.06
MDL Number MFCD00060659
InChI Key ZLMKEENUYIUKKC-UHFFFAOYSA-N
Synonym 5-iodo-m-xylene, 1,3-dimethyl-5-iodobenzene, benzene, 1-iodo-3,5-dimethyl, 3,5-dimethyliodobenzene, pubchem3872, acmc-1ccdf, 3,5-dimethyl-1-iodobenzene, ksc422q9l, 1-iodo-3,5-dimethylbenzene, #
PubChem CID 140924
IUPAC Name 1-iodo-3,5-dimethylbenzene
SMILES CC1=CC(I)=CC(C)=C1

Specifications

Color Yellow
Quantity 25 g
Physical Form Clear Liquid at 20°C
Boiling Point 229°C
Formula Weight 232.06
Percent Purity ≥98.0% (GC)
Grade Not Available
Chemical Name or Material 5-Iodo-m-xylene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.