Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Iodo-o-toluic Acid 98.0+%, TCI America™
SDP

Catalog No. I09165G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I09165G 5 g
I091625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I09165G Supplier TCI America Supplier No. I09165G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 54811-38-0
Molecular Formula C8H7IO2
Molecular Weight (g/mol) 262.046
MDL Number MFCD01570258
InChI Key WUBHOZQZSHGUFI-UHFFFAOYSA-N
Synonym 5-Iodo-2-methylbenzoic Acid
PubChem CID 621745
IUPAC Name 5-iodo-2-methylbenzoic acid
SMILES CC1=C(C=C(C=C1)I)C(=O)O

Specifications

Melting Point 181°C
Color White
Quantity 5 g
Formula Weight 262.05
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Iodo-o-toluic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.